Learn More
Fine Science Tools Dual Clasping Stainless Steel Sterilization Container
SDP
Catalog No. NC9013646
Encompass
Product: Dual Clasping Stainless Steel Sterilization Container - 30 x 14 x 4cm/ Dimensions: 30 x 14 x 4cm/ Alloy / Material: Stainless Steel/ Autoclave Safe: Yes/ Unit: Each
Catalog No. NC9013646 Supplier Fine Science Tools Supplier No. 2085000
Product: Dual Clasping Stainless Steel Sterilization Container - 30 x 14 x 4cm/ Dimensions: 30 x 14 x 4cm/ Alloy / Material: Stainless Steel/ Autoclave Safe: Yes/ Unit: Each