Learn More
PHCbi Sliding Drawer Racks for PHCbi Upright Freezers
Catalog No. NC3073661
Encompass_Preferred
These sliding drawer racks hold 2" and 3" fiberboard boxes and are for use with PHCbi upright freezers.
Catalog No. NC3073661 Supplier PHCbi Supplier No. SUR624P
These sliding drawer racks hold 2" and 3" fiberboard boxes and are for use with PHCbi upright freezers.