Learn More
Corning™ Stainless Steel Support Rod for Hot Plate Stirrers Produced in China
Catalog No. p-200104862 Shop All Corning Products
Change view
Click to view available options
Length (English) Rod:
12 in.
18 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Rod
08-757-302 12 in.
08-757-303 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-757-302 Supplier Corning™ Supplier No. 400084CH
Only null left
Add to Cart
Add to Cart
Stainless steel support rod designed for hot plate stirrers produced in China.
Corning offers a stainless steel, two piece steel support rod that consists of two rods that screw together.
  • Screws into the base of Corning hot plate and stirrers
TRUSTED_SUSTAINABILITY
Specifications
Length (English) Rod 12 in.
Diameter (English) Rod 5/16 in.
Product Type Stirrer Support Rod
For Use With (Equipment) Corning PC-200/210/220 Series Hot Plate/Stirrers
Quantity 1 Ea.