Learn More
Perkin Elmer US LLC Stainless Steel TD tubes Un-Conditioned
Catalog No. 502104721
Encompass_Preferred
Sorbent: Soil Vapor Intrusion Our new multi-bed construction extends the hydrocarbon range past naphthalene while retaining the lighter components, enabling larger sample volumes, hence, enhancing detection limits. Unique design that meets the challenges and criteria of the EPA regulations for air monitoring. Enhanced recoveries, even of highly contaminated sites From chloromethane through diesel range hydrocarbons After the analysis, tubes are clean and ready for re-sampling reducing costs.
Catalog No. 50-210-4721 Supplier Perkin Elmer US LLC Supplier No. N9306278
Sorbent: Soil Vapor Intrusion Our new multi-bed construction extends the hydrocarbon range past naphthalene while retaining the lighter components, enabling larger sample volumes, hence, enhancing detection limits. Unique design that meets the challenges and criteria of the EPA regulations for air monitoring. Enhanced recoveries, even of highly contaminated sites From chloromethane through diesel range hydrocarbons After the analysis, tubes are clean and ready for re-sampling reducing costs.