Learn More
Perkin Elmer US LLC Stainless Steel Triple Filter Kit - Oxygen/Moisture/Hydrocarbon, 0.125 in
Supplier:  Perkin Elmer US LLC N9306831
Encompass_Preferred
The triple combination filter kit is ideal for purifying GC/MS carrier gases. It contains oxygen, moisture and hydrocarbon scrubbers in one easy to change economical cartridge. Includes: (1) one-position base plate with 1/8 inch stainless steel inlet/outlet fittings (1) Oxygen/Moisture/Hydrocarbon trap
Catalog No. 50-211-3662