Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for 25-Place Slide Boxes, 7x5, 35 Box Capacity
SDP
Catalog No. 503028054
Encompass_Preferred
For 25- Place Slide Boxes The UFS-25-75 standard upright freezer racks for storage of 25-place slide boxes, features 7 boxes deep by 5 boxes high configuration for a total storage of 35 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with 25-place slide boxes. Dimensions (DxWxH): 26 3/8 x 5 11/16 x 7 11/16 inches (670 x 145 x 195mm)Compartment Dimensions: 3 5/8 x 5 5/8 x 1 1/2 inches (92 x 143 x 38mm)
Catalog No. 50-302-8054 Supplier Alkali Scientific Supplier No. UFS2575
For 25- Place Slide Boxes The UFS-25-75 standard upright freezer racks for storage of 25-place slide boxes, features 7 boxes deep by 5 boxes high configuration for a total storage of 35 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with 25-place slide boxes. Dimensions (DxWxH): 26 3/8 x 5 11/16 x 7 11/16 inches (670 x 145 x 195mm)Compartment Dimensions: 3 5/8 x 5 5/8 x 1 1/2 inches (92 x 143 x 38mm)