Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for 25-Place Slide Boxes, 6x4, 24 Box Capacity
SDP
Catalog No. 503028057
Encompass_Preferred
For 25- Place Slide Boxes The UFS-25-64 standard upright freezer racks for storage of 25-place slide boxes, features 6 boxes deep by 4 boxes high configuration for a total storage of 24 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with 25-place slide boxes. Dimensions (DxWxH): 22 11/16 x 5 11/16 x 6 3/16 inches (576.5 x 145 x 156.5mm)Compartment Dimensions: 3 5/8 x 5 5/8 x 1 1/2 inches (92 x 143 x 38mm)
Catalog No. 50-302-8057 Supplier Alkali Scientific Supplier No. UFS2564
For 25- Place Slide Boxes The UFS-25-64 standard upright freezer racks for storage of 25-place slide boxes, features 6 boxes deep by 4 boxes high configuration for a total storage of 24 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with 25-place slide boxes. Dimensions (DxWxH): 22 11/16 x 5 11/16 x 6 3/16 inches (576.5 x 145 x 156.5mm)Compartment Dimensions: 3 5/8 x 5 5/8 x 1 1/2 inches (92 x 143 x 38mm)