Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Drawer Rack for MATRIX Boxes, FluidX Boxes & Micronic Racks ,5x3 configuration, 15 Box Capacity
SDP
Catalog No. 503028442
Encompass_Preferred
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 2 1/8″- 2 7/16″ (54.8-61.8mm) The UFDMX-532 upright freezer drawer racks are designed to safely store a total of (15) 96 Well MATRIX Boxes, FluidX Boxes or Micronic Racks in an upright laboratory freezer. These racks are made out of corrosion resistant stainless steel. Upright freezer drawer racks include handles and identification card holders. Boxes are not included with purchase. Dimensions L x H x W: 17 7/8 x 7 15/16 x 5 7/16 inch ( 454 x 202 x 138mm)Compartment Size l x h x W: 3 7/16 x 2 7/16 x 5 1/16 inch (88 x 62.4 x 129mm)
Catalog No. 50-302-8442 Supplier Alkali Scientific Supplier No. UFDMX532
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 2 1/8″- 2 7/16″ (54.8-61.8mm) The UFDMX-532 upright freezer drawer racks are designed to safely store a total of (15) 96 Well MATRIX Boxes, FluidX Boxes or Micronic Racks in an upright laboratory freezer. These racks are made out of corrosion resistant stainless steel. Upright freezer drawer racks include handles and identification card holders. Boxes are not included with purchase. Dimensions L x H x W: 17 7/8 x 7 15/16 x 5 7/16 inch ( 454 x 202 x 138mm)Compartment Size l x h x W: 3 7/16 x 2 7/16 x 5 1/16 inch (88 x 62.4 x 129mm)