Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Racks for 15mL & 50mL Tube Boxes, 3 x 2 Configuration, 6 Box Capacity
SDP
Catalog No. 503028533
Encompass_Preferred
Stainless steel upright freezer rack for storage of 15ml & 50ml Centrifuge Tube Boxes is designed to fit a total of 8 boxes. These freezer racks come standard with Locking Rods and are constructed of corrosion resistant stainless steel. Security lock devices & storage boxes are also available for each rack though ordered separately. *Tubes and boxes not included Dimensions (DxWxH): 18 13/16 x 5 7/8 x 10 3/16 inches (478 x 149 x 259mm)Compartment Dimensions: 6  x 5 13/16 x 5 1/16 inches (152 x 147.5 x 128mm)  
Catalog No. 50-302-8533 Supplier Alkali Scientific Supplier No. UFLBR32
Stainless steel upright freezer rack for storage of 15ml & 50ml Centrifuge Tube Boxes is designed to fit a total of 8 boxes. These freezer racks come standard with Locking Rods and are constructed of corrosion resistant stainless steel. Security lock devices & storage boxes are also available for each rack though ordered separately. *Tubes and boxes not included Dimensions (DxWxH): 18 13/16 x 5 7/8 x 10 3/16 inches (478 x 149 x 259mm)Compartment Dimensions: 6  x 5 13/16 x 5 1/16 inches (152 x 147.5 x 128mm)  