Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for 96-Well Microtube Boxes, 4 x 3 Configuration, 12 Box Capacity
SDP
Catalog No. 503028906
Encompass_Preferred
For 96-Well Microtube BoxesFor Speciality Tube Racks (Matrix, Abgene, Fluidx, Micronic)- H: 5/8″- 2 1/8″ (17.3-53.5mm)Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.   Racks are for 96-Well Microtube Boxes.  Each rack compartment holds a single 96-Well Microtube Box.  Includes locking rod.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Dimensions (DxWxH): 15 3/16 x 5 1/2 x 6 13/16 inches (385 x 140 x 173mm)Compartment Dimensions: 3 9/16 x 2 3/16 x 5 3/16inch (91 x 55 x 130.7mm)
Catalog No. 50-302-8906 Supplier Alkali Scientific Supplier No. FR5219
For 96-Well Microtube BoxesFor Speciality Tube Racks (Matrix, Abgene, Fluidx, Micronic)- H: 5/8″- 2 1/8″ (17.3-53.5mm)Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.   Racks are for 96-Well Microtube Boxes.  Each rack compartment holds a single 96-Well Microtube Box.  Includes locking rod.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Dimensions (DxWxH): 15 3/16 x 5 1/2 x 6 13/16 inches (385 x 140 x 173mm)Compartment Dimensions: 3 9/16 x 2 3/16 x 5 3/16inch (91 x 55 x 130.7mm)