Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for 0.2ml Tube 96-Well PCR Boxes 6 x 8 Configuration, 56 Box Capacity
SDP
Catalog No. 503027569
Encompass_Preferred
For 0.2 ml Tube 96-Well PCR Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.  Upright Freezer Racks for 0.2ml Tube 96-well PCR Boxes.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Features: Constructed of corrosion resistant stainless steel Locking rods are included Security lock devices are availble for each rack Sturdy handle design Dimensions (DxWxH): 22 1/2 x 12 1/16 x 5 1/2inch (571 x 306 x 140mm)Compartment Dimensions: 3 9/16 x 1 7/16 x 5 3/16 inch (91 x 37 x 130.5mm)
Catalog No. 50-302-7569 Supplier Alkali Scientific Supplier No. UFPCR68L
For 0.2 ml Tube 96-Well PCR Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.  Upright Freezer Racks for 0.2ml Tube 96-well PCR Boxes.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Features: Constructed of corrosion resistant stainless steel Locking rods are included Security lock devices are availble for each rack Sturdy handle design Dimensions (DxWxH): 22 1/2 x 12 1/16 x 5 1/2inch (571 x 306 x 140mm)Compartment Dimensions: 3 9/16 x 1 7/16 x 5 3/16 inch (91 x 37 x 130.5mm)