Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for 80 Place Tube Boxes, 8 x 5 Configuration, 40 Box Capacity
SDP
Catalog No. 503026704
Encompass_Preferred
For 80-Place Tube Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space. Handles at both ends accommodate freezers with right or left handed doors.  Upright Freezer Racks for 80 Place Tube Boxes.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Dimensions (DxWxH): 23 1/8 x 9 3/16 x 6 11/16 inches (588 x 234 x 170mm)Compartment Dimensions: 2 3/4 x 9 1/8 x 1 5/16 inches (70 x 231 x 33mm)
Catalog No. 50-302-6704 Supplier Alkali Scientific Supplier No. UT8085
For 80-Place Tube Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space. Handles at both ends accommodate freezers with right or left handed doors.  Upright Freezer Racks for 80 Place Tube Boxes.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Dimensions (DxWxH): 23 1/8 x 9 3/16 x 6 11/16 inches (588 x 234 x 170mm)Compartment Dimensions: 2 3/4 x 9 1/8 x 1 5/16 inches (70 x 231 x 33mm)