Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Drawer Rack for MATRIX Boxes, FluidX Boxes & Micronic Racks – 16 Box Capacity
SDP
Catalog No. 503027935
Encompass_Preferred
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 2 1/8″- 2 7/16″ (54.8-61.8mm) The UFDMX-442 upright freezer drawer racks are designed to safely store a total of (16) 96 Well MATRIX Boxes, FluidX Boxes or Micronic Racks in an upright laboratory freezer. These racks are made out of corrosion resistant stainless steel. Upright freezer drawer racks include handles and identification card holders. Boxes are not included with purchase. Dimensions L x H x W: 14 3/8 x 10 9/16 x 5 7/16 inch ( 365 x 268.5 x 138mm)Compartment Size l x h x W: 3 7/16 x 2 7/16 x 5 1/16 inch (88 x 62.4 x 129mm)
Catalog No. 50-302-7935 Supplier Alkali Scientific Supplier No. UFDMX442
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 2 1/8″- 2 7/16″ (54.8-61.8mm) The UFDMX-442 upright freezer drawer racks are designed to safely store a total of (16) 96 Well MATRIX Boxes, FluidX Boxes or Micronic Racks in an upright laboratory freezer. These racks are made out of corrosion resistant stainless steel. Upright freezer drawer racks include handles and identification card holders. Boxes are not included with purchase. Dimensions L x H x W: 14 3/8 x 10 9/16 x 5 7/16 inch ( 365 x 268.5 x 138mm)Compartment Size l x h x W: 3 7/16 x 2 7/16 x 5 1/16 inch (88 x 62.4 x 129mm)