Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for 25-Place Slide Boxes, 18 Box Capacity
SDP
Catalog No. 503028009
Encompass_Preferred
For 25- Place Slide Boxes The UFS-25-36 standard upright freezer racks for storage of 25-place slide boxes, features 3 boxes deep by 6 boxes high configuration for a total storage of 18 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with 25-place slide boxes. Dimensions L x H x W: 11 1/2 x 10 3/16 x 6 inch (291.5 x 259 x 152 mm)Compartment Dimensions l x h x w: 3 11/16 x 1 1/2 x 5 9/16 inch (92.5 x 38 x 141mm)
Catalog No. 50-302-8009 Supplier Alkali Scientific Supplier No. UFS2536
For 25- Place Slide Boxes The UFS-25-36 standard upright freezer racks for storage of 25-place slide boxes, features 3 boxes deep by 6 boxes high configuration for a total storage of 18 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with 25-place slide boxes. Dimensions L x H x W: 11 1/2 x 10 3/16 x 6 inch (291.5 x 259 x 152 mm)Compartment Dimensions l x h x w: 3 11/16 x 1 1/2 x 5 9/16 inch (92.5 x 38 x 141mm)