Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for Plasma Boxes, 3x6 configuration, 18 Box Capacity
SDP
Catalog No. 503028041
Encompass_Preferred
For Plasma Boxes Our freezer racks feature pharmaceutical grade stainless steel, double welds for long life and smooth edges for your safety and comfort.  Made of corrosion resistant stainless steel Locking rods are included Dimension L x H x W: 22 3/8 x 12 5/8 x 5 1/2 inch (568 x 321 x 140mm)Compartment Dimension l x h x w: 7 3/16 x 2 1/16 x 5 3/8 inch ( 183 x 52.5 x 136.7mm)
Catalog No. 50-302-8041 Supplier Alkali Scientific Supplier No. UFP36
For Plasma Boxes Our freezer racks feature pharmaceutical grade stainless steel, double welds for long life and smooth edges for your safety and comfort.  Made of corrosion resistant stainless steel Locking rods are included Dimension L x H x W: 22 3/8 x 12 5/8 x 5 1/2 inch (568 x 321 x 140mm)Compartment Dimension l x h x w: 7 3/16 x 2 1/16 x 5 3/8 inch ( 183 x 52.5 x 136.7mm)