Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for SBS Formatted Boxes (Micronic, Fluidx), 4x3 configuration, 12 Box Capacity
SDP
Catalog No. 503026692
Encompass_Preferred
For Speciality Tube Racks (Micronic, Fluidx)- H: 4-4 1/4″ (101.5-107.82mm) Our freezer racks feature pharmaceutical grade stainless steel, double welds for long life and smooth edges for your safety and comfort.  Made of corrosion resistant stainless steel Locking rods are included Dimensions (DxWxH): 14 11/16 x 5 7/16 x 13 3/8 inches (373 x 138 x 340mm)Compartment Dimensions: 3 7/16 x 5 1/8 x 4 3/8 inches (88 x 130 x 110.5mm)
Catalog No. 50-302-6692 Supplier Alkali Scientific Supplier No. UFMR43
For Speciality Tube Racks (Micronic, Fluidx)- H: 4-4 1/4″ (101.5-107.82mm) Our freezer racks feature pharmaceutical grade stainless steel, double welds for long life and smooth edges for your safety and comfort.  Made of corrosion resistant stainless steel Locking rods are included Dimensions (DxWxH): 14 11/16 x 5 7/16 x 13 3/8 inches (373 x 138 x 340mm)Compartment Dimensions: 3 7/16 x 5 1/8 x 4 3/8 inches (88 x 130 x 110.5mm)