Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for 96-Well Flipper Reversible Boxes, 5 x 6 Configuration, 30 Box Capacity
SDP
Catalog No. 503027902
Encompass_Preferred
For 96-Well Flipper Reversible Boxes Our freezer racks feature pharmaceutical grade stainless steel, double welds for long life and smooth edges for your safety and comfort.  Made of corrosion resistant stainless steel Dimensions (DxWxH): 24 13/16 x 8 3/4 x 12 5/8 inches (630 x 222 x 321mm)Compartment Dimensions: 4 3/4 x 8 5/8 x 2 1/16 inches (121 x 219 x 52.6mm)  
Catalog No. 50-302-7902 Supplier Alkali Scientific Supplier No. UFR56
For 96-Well Flipper Reversible Boxes Our freezer racks feature pharmaceutical grade stainless steel, double welds for long life and smooth edges for your safety and comfort.  Made of corrosion resistant stainless steel Dimensions (DxWxH): 24 13/16 x 8 3/4 x 12 5/8 inches (630 x 222 x 321mm)Compartment Dimensions: 4 3/4 x 8 5/8 x 2 1/16 inches (121 x 219 x 52.6mm)  