Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for 25-Place Slide Boxes, 36 Box Capacity
SDP
Supplier:  Alkali Scientific UFS2566
Encompass_Preferred
Top of Form For 25- Place Slide Boxes The UFS-25-66 standard upright freezer racks for storage of 25-place slide boxes, features a 6 boxes deep by 6 boxes high configuration for a total storage of 36 boxes per rack. These upright freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rack does not come standard with 25-place slide boxes. Dimensions (DxWxH): 22 11/16 x 5 11/16 x 9 3/16 inches (576.5 x 145 x 233.5mm)Compartment Dimensions: 3 5/8 x 5 5/8 x 1 1/2 inches (92 x 143 x 38mm)
Catalog No. 50-302-8122