Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for MATRIX Boxes, FluidX Boxes & Micronic Racks – 3 Boxes Deep x 5 Boxes High
SDP
Catalog No. 503028544
Encompass_Preferred
For Speciality Tube Racks (Matrix, Abgene, Fluidx, Micronic)- H: 2 5/18″- 2 7/16″ (54.8-61.8mm) The UFMX-352 upright freezer racks for 96 Well MATRIX boxes, FluidX Boxes & Micronic Racks are designed to safely store a total of (15) boxes in an upright laboratory freezer. These freezer racks are built out of high-quality stainless steel and offer convenient storage of samples in an organized format. These upright freezer racks include handles and locking rods for added security. Boxes are not included and must be purchased separately. Dimensions (DxWxH): 11 1/8 x 5 7/16 x 12 5/8 inches (283 x 138 x 321mm)Compartment Dimensions: 3 7/16 x 5 1/8 x 2 1/2inches (88 x 130 x 62.5mm)
Catalog No. 50-302-8544 Supplier Alkali Scientific Supplier No. UFMX352
For Speciality Tube Racks (Matrix, Abgene, Fluidx, Micronic)- H: 2 5/18″- 2 7/16″ (54.8-61.8mm) The UFMX-352 upright freezer racks for 96 Well MATRIX boxes, FluidX Boxes & Micronic Racks are designed to safely store a total of (15) boxes in an upright laboratory freezer. These freezer racks are built out of high-quality stainless steel and offer convenient storage of samples in an organized format. These upright freezer racks include handles and locking rods for added security. Boxes are not included and must be purchased separately. Dimensions (DxWxH): 11 1/8 x 5 7/16 x 12 5/8 inches (283 x 138 x 321mm)Compartment Dimensions: 3 7/16 x 5 1/8 x 2 1/2inches (88 x 130 x 62.5mm)