Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for MATRIX Boxes, FluidX Boxes & Micronic Racks – 6 Boxes Deep x 3 Boxes High
SDP
Catalog No. 503028503
Encompass_Preferred
The UFMX-632 upright freezer racks for 96 Well MATRIX boxes, FluidX Boxes & Micronic Racks are designed to safely store a total of (18) boxes in an upright laboratory freezer. These freezer racks are built out of high-quality stainless steel and offer convenient storage of samples in an organized format. These upright freezer racks include handles and locking rods for added security. Boxes are not included and must be purchased separately.
Catalog No. 50-302-8503 Supplier Alkali Scientific Supplier No. UFMX632
The UFMX-632 upright freezer racks for 96 Well MATRIX boxes, FluidX Boxes & Micronic Racks are designed to safely store a total of (18) boxes in an upright laboratory freezer. These freezer racks are built out of high-quality stainless steel and offer convenient storage of samples in an organized format. These upright freezer racks include handles and locking rods for added security. Boxes are not included and must be purchased separately.