Learn More
Lakeside Manufacturing Inc Stainless Steel Utility Cart, 3 Shelf: 15-1/2" x 24", 300-Lb. Capacity
Supplier:  Lakeside Manufacturing Inc 311
Encompass_Preferred
Stainless Steel Utility Carts by Lakeside Manufacturing are ideal for hauling and restocking medical supplies, surgical equipment set-up, light duty transport and storage. The Lakeside 311 Stainless Steel Utility Cart with (3) shelves measuring 15-1/2" x 24" has a 300-Lb. weight capacity. This cart's bumpers on its legs and handles help protect walls and furniture, while its all-welded angle leg design provides durability. This utility cart's sound deadening material under its shelves restricts vibration and reduces noise. This medical utility cart on wheels has a reinforced, hemmed front that adds strength where the cart is most vulnerable to stress. Its stainless steel construction is easy to clean and sanitize. Unitized electronic welding is used to fasten the 311's shelves and handle to the frame. Ideal for transporting light duty loads up to 300-Lb. in clinical environments. Endless variations and feature combination. Superior lead times. Made in the USA.
Catalog No. NC1884696