Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Vertical Rack for SBS formatted boxes, Holds 13 boxes
SDP
Catalog No. 503029411
Encompass_Preferred
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 1 1/4″- 1 1/2″ (31.7-38.7mm) Dimensions L x H x W: 21 15/16 x 3 5/8 x 5 1/2 inch (557 x 92 x 140mm)Compartment Size l x h x W: 1 3/16 x 3 7/16 x 5 3/16 inch (30 x 88 x 132mm)
Catalog No. 50-302-9411 Supplier Alkali Scientific Supplier No. CFML13
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 1 1/4″- 1 1/2″ (31.7-38.7mm) Dimensions L x H x W: 21 15/16 x 3 5/8 x 5 1/2 inch (557 x 92 x 140mm)Compartment Size l x h x W: 1 3/16 x 3 7/16 x 5 3/16 inch (30 x 88 x 132mm)