Learn More
Troemner™ Stainless-Steel Weight Sets
SCI_ED
Catalog No. S40203A Shop All Troemner Products
Change view
Click to view available options
Packaging Quantity:
20
21
Quantity:
20/Set
21/Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity Quantity
S40203A 21 21/Set
S40202A 20 20/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S40203A Supplier Troemner™ Supplier No. 30390968
Add to Cart
Add to Cart
Economical Stainless Steel Class 7 Weight Set with No Certificate
Weights are adjusted to tolerances specified by ANSI™/ASTM™ Class 7. Each set comes complete with a plastic storage container.
  • Fractional masses of 200mg and over are made of stainless steel
  • Fractional masses below 200mg are made of aluminum
  • Sets with fractional masses include forceps
TRUSTED_SUSTAINABILITY
Specifications
Product Type Metric Economical Weight Set
Classification ASTM Class 7
Material Stainless Steel
Weight (Metric) 1 mg to 100 g
Mass Various
Includes 303 Stainless Steel cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, 303 Stainless Steel sheet weight: 500mg, 2 x 200mg, Aluminum sheet weight: 100mg, 50mg, 2 x 20mg, 10mg, 5mg, 2 x 2mg, 1mg, Plastic case
Quantity 21/Set
Packaging Quantity 21