Learn More
MilliporeSigma™ Supelco™ Stainless Steel Weights
Catalog No. 111005167 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
11-100-5167 4/Pk.
1 options
Catalog No. 11-100-5167 Supplier MilliporeSigma™ Supelco™ Supplier No. 57278
Add to Cart
Add to Cart
Stainless steel weights for Visiprep large volume are components that are fitted at one end of visiprep large volume samplers, which also consists of 1/8-inch polytetrafluoroethylene (PTFE) tubing with a screw fitting at the other end.
TRUSTED_SUSTAINABILITY
Specifications
Description Weight fitting for tubing 1/8 in. O.D.
For Use With (Application) Food and Beverages
For Use With (Equipment) Visiprep Large Volume Sampler
Product Line Visiprep
Type SPE Accessory
Material Stainless steel
Quantity 4/Pk.
Visiprep is a trademark of Sigma-Aldrich Co. LLC