Learn More
TA Instruments Stainless Steel, Wide Gap Vaned Rotor with Heat Break, Rotor Radius 7.5 mm, Rotor Height 38 mm, Number of Vanes 4, for Use with Standard 15 mm Radius Cup, HR X0, Smart Swap 2
Catalog No. 5000580645 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Smart Swap 2, Stainless Steel, Wide Gap Vaned Rotor With Heat Break has a rotor radius of 7.5 mm, rotor height of 38 mm, and 4 vanes. The rotor is designed for use with the standard 15 mm radius cup.
Catalog No. 50-005-80645 Supplier TA Instruments Supplier No. 546027.941
Waters TA Instruments HR X0, Smart Swap 2, Stainless Steel, Wide Gap Vaned Rotor With Heat Break has a rotor radius of 7.5 mm, rotor height of 38 mm, and 4 vanes. The rotor is designed for use with the standard 15 mm radius cup.