Learn More
Analytical Sales & Services Stainless Steel ZDV Union
Catalog No. 5000805055
Encompass
Ultra High Pressure SS Union for 1/16" OD Tubing\n\nQty/Pkg: EACH
Catalog No. 50-008-05055 Supplier Analytical Sales & Services Supplier No. 84161
Ultra High Pressure SS Union for 1/16" OD Tubing\n\nQty/Pkg: EACH