Learn More
Analytical Sales & Services Stainless Steel ZDV Union, SSI Compatible
Catalog No. 5000806477
Encompass
SSI Compatible ZDV Union\n\n.020" Thru-Hole\n\nQty/Pkg: EACH
Catalog No. 50-008-06477 Supplier Analytical Sales & Services Supplier No. 64352
SSI Compatible ZDV Union\n\n.020" Thru-Hole\n\nQty/Pkg: EACH