Learn More
Sklar Instruments Disposable Stainless Steel Surgical Blades, Sklar 10, BX100.
SDP
Catalog No. 501094389 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Stainless Steel Surgical Blades, Sklar, #10. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #10 is most commonly used for large, straight incisions. It has a curved cutting edge. These disposable blades are available in bulk quantities of 100 per box.
Catalog No. 50-109-4389 Supplier Sklar Instruments Supplier No. 063100
Disposable Stainless Steel Surgical Blades, Sklar, #10. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #10 is most commonly used for large, straight incisions. It has a curved cutting edge. These disposable blades are available in bulk quantities of 100 per box.