Learn More
Sklar Instruments Disposable Stainless Steel Surgical Blades, Sklar 11, BX100.
SDP
Catalog No. 501094390 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Stainless Steel Surgical Blades, Sklar, #11. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #11 is most commonly used for puncture incisions or to open an artery. It has an angled cutting edge that leads to an extremely sharp point. Care should be taken to avoid inadvertently passing the blade too deep into the skin. These disposable blades are available in bulk quantities of 100 per box.
Catalog No. 50-109-4390 Supplier Sklar Instruments Supplier No. 063101
Disposable Stainless Steel Surgical Blades, Sklar, #11. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #11 is most commonly used for puncture incisions or to open an artery. It has an angled cutting edge that leads to an extremely sharp point. Care should be taken to avoid inadvertently passing the blade too deep into the skin. These disposable blades are available in bulk quantities of 100 per box.