Learn More
Sklar Instruments Disposable Stainless Steel Surgical Blades, Sklar 12, BX100.
SDP
Catalog No. 501094391 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Stainless Steel Surgical Blades, Sklar, #12. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #12 is a small, pointed, crescent shaped blade sharpened along the inside edge of the curve. These disposable blades are available in bulk quantities of 100 per box.
Catalog No. 50-109-4391 Supplier Sklar Instruments Supplier No. 063102
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disposable Stainless Steel Surgical Blades, Sklar, #12. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #12 is a small, pointed, crescent shaped blade sharpened along the inside edge of the curve. These disposable blades are available in bulk quantities of 100 per box.