Learn More
Sklar Instruments Disposable Stainless Steel Surgical Blades, Sklar #21. Sklar Disposable, Sterile, Stainless Steel Surgical Blade #21, BX100.
SDP
Catalog No. 501094394 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Stainless Steel Surgical Blades, Sklar, #21. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #21 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #21 blade is smaller than the #22 blade and larger than the #20 blade and is compatible with the #4 series and larger handles. This blade is one of the most traditional, commonly used blades and often used for making small incisions in skin and muscle. These disposable blades are available in bulk quantities of 100 per box.
Catalog No. 50-109-4394 Supplier Sklar Instruments Supplier No. 063105
Disposable Stainless Steel Surgical Blades, Sklar, #21. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #21 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #21 blade is smaller than the #22 blade and larger than the #20 blade and is compatible with the #4 series and larger handles. This blade is one of the most traditional, commonly used blades and often used for making small incisions in skin and muscle. These disposable blades are available in bulk quantities of 100 per box.