Learn More
Uline Stainless Steel Wire Shelving Unit - 72 x 24 x 72""""
Catalog No. NC1151220
Encompass
72 x 24 x 72"""" freestanding wire shelving unit, stainless steel (type 304) allows air circulation and visibility of stored items, shelf capacity = 650lbs, 4 shelves
Catalog No. NC1151220 Supplier Uline Supplier No. H4800
72 x 24 x 72"""" freestanding wire shelving unit, stainless steel (type 304) allows air circulation and visibility of stored items, shelf capacity = 650lbs, 4 shelves