Learn More
Thermo Scientific Chemicals Stains-All, 95%
Description
Stains-all is an ideal dye for the visualization and identification of proteins on polyacrylamide gels because it differentiates between highly acidic proteins which stain blue and less acidic proteins which stain pink. When employed on polyacrylamide gels, the intensity of Stains-all can be increased by the addition of silver nitrate. It is a useful nucleic acid and protein stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1g |
| Storage Requirements | 2° to 8°C |
| Format | Powder |
| For Use With (Application) | Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids |
| Color | Dark Gray or Green |
| Formula Weight | 559.58g/mol |
| Sensitivity | Light sensitive |
| Product Type | Hematology Stain |
| Assay | 0.95 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.