Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Stains-All, 95%

Catalog No. AAJ6263006
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6263006 5g
AAJ6263003 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6263006 Supplier Thermo Scientific Chemicals Supplier No. J6263006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids | CAS: 7423-31-6 | C30H27BrN2S2, C30H27BrN2S2 | 559.58 g/mol

Stains-all is an ideal dye for the visualization and identification of proteins on polyacrylamide gels because it differentiates between highly acidic proteins which stain blue and less acidic proteins which stain pink. When employed on polyacrylamide gels, the intensity of Stains-all can be increased by the addition of silver nitrate. It is a useful nucleic acid and protein stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-31-6
Molecular Formula C30H27BrN2S2

Specifications

Quantity 5g
Storage Requirements 2° to 8°C
Format Powder
For Use With (Application) Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids
Color Dark Gray or Green
Formula Weight 559.58g/mol
Sensitivity Light sensitive
Product Type Hematology Stain
Assay 0.95

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.