Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Stains-All, 96%

Catalog No. AAH3212706
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3212706 5g
AAH3212703 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3212706 Supplier Thermo Scientific Chemicals Supplier No. H3212706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7423-31-6 | C30H27BrN2S2

General protein and nucleic acid stain, which stains different colors on bands of RNA (bluish purple), DNA (blue) and proteins (red)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-31-6
Molecular Formula C30H27BrN2S2
Synonym 1-Ethyl-2-[(E)-3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide; 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine

Specifications

Formula Weight 559.58g/mol
Quantity 5g
Product Type Stain
Assay 0.96
Is Stains-All, 96% suitable for RNA purification by SDS-PAGE?

Yes, Stains-All, 96% is a cationic cyanine dye, and it is possible to stain RNA with it. Gel staining should be performed in the dark, as cyanine dyes can be excited with visible light to produce ROS, leading to damage of biomolecules.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.