Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Stains-all, 97%, pure

Catalog No. AC213512500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213512500 250 mg Glass bottle
AC213510010 1 g Glass bottle
AC213510050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC213512500 Supplier Thermo Scientific Chemicals Supplier No. 213512500
  • $66.75
    $54.65 / Each of 1

    Save $12.10

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 7423-31-6 | C30H28BrN2S2+ | 560.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-31-6
Molecular Formula C30H28BrN2S2+
Molecular Weight (g/mol) 560.592
MDL Number MFCD00078788
InChI Key MPBRYMWMMKKRGC-UHFFFAOYSA-N
Synonym 1-Ethyl-2-[3-(1-ethylnaphtho[1, 2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1, 2-d]thiazolium bromide
PubChem CID 123134119
IUPAC Name (2E)-1-ethyl-2-[(E)-3-(1-ethylbenzo[e][1,3]benzothiazol-1-ium-2-yl)-2-methylprop-2-enylidene]benzo[e][1,3]benzothiazole;hydrobromide
SMILES CCN1C(=CC(=CC2=[N+](C3=C(S2)C=CC4=CC=CC=C43)CC)C)SC5=C1C6=CC=CC=C6C=C5.Br

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
UV Spectrometry Complies
Beilstein 27, IV, 8619
Color Gray to Purple
Quantity 250 mg
Physical Form Powder
Formula Weight 559.59
Percent Purity 97%
Grade Pure
Chemical Name or Material Stains-all, 97%
Show More Show Less
Hazard Category LIGHT SENSITIVE
EINECSNumber 231-047-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.