Learn More
Ethos Biosciences Lugol's Iodine Concentrate, 4oz, Ethos
Catalog No. 5000574270
Encompass_Preferred
Lugol’s Iodine Concentrate is a high-strength iodine solution designed for microbiology, parasitology, cytology, and general laboratory applications. Intended for dilution to working strength, it is commonly used as a staining and contrast reagent to enhance visualization of cellular structures, microorganisms, and parasitic elements during microscopic examination. The concentrate provides consistent composition for reliable preparation of working solutions and reproducible staining results. It is also suitable for general laboratory reagent preparation and analytical procedures. Supplied in chemically compatible containers, Lugol’s Iodine Concentrate is appropriate for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74270 Supplier Ethos Biosciences Supplier No. 658404
Lugol’s Iodine Concentrate is a high-strength iodine solution designed for microbiology, parasitology, cytology, and general laboratory applications. Intended for dilution to working strength, it is commonly used as a staining and contrast reagent to enhance visualization of cellular structures, microorganisms, and parasitic elements during microscopic examination. The concentrate provides consistent composition for reliable preparation of working solutions and reproducible staining results. It is also suitable for general laboratory reagent preparation and analytical procedures. Supplied in chemically compatible containers, Lugol’s Iodine Concentrate is appropriate for clinical, research, and educational laboratory environments.