Learn More
Ethos Biosciences Lugol's Iodine Working Solution, 16oz, Ethos
Catalog No. 5000574126
Encompass_Preferred
Lugol’s Iodine Working Solution is a ready-to-use iodine reagent suitable for microbiology, parasitology, cytology, and general laboratory applications. Commonly used as a staining and contrast agent, it enhances the visualization of cellular structures, microorganisms, and parasitic elements during microscopic examination. The working-strength formulation provides consistent color development for reliable slide preparation and evaluation. It is also suitable for general laboratory staining and reagent preparation tasks. Supplied in chemically compatible containers, Lugol’s Iodine Working Solution is appropriate for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74126 Supplier Ethos Biosciences Supplier No. 658316
Lugol’s Iodine Working Solution is a ready-to-use iodine reagent suitable for microbiology, parasitology, cytology, and general laboratory applications. Commonly used as a staining and contrast agent, it enhances the visualization of cellular structures, microorganisms, and parasitic elements during microscopic examination. The working-strength formulation provides consistent color development for reliable slide preparation and evaluation. It is also suitable for general laboratory staining and reagent preparation tasks. Supplied in chemically compatible containers, Lugol’s Iodine Working Solution is appropriate for clinical, research, and educational laboratory environments.