Learn More
StatLab StainSafe Xylene Resistant Slide Labels
SDP
Catalog No. 21010368
Change view
Click to view available options
Product Type:
Slide Labels
Catalog No. Product Type
21-010-368 Slide Labels
1 options
Catalog No. 21-010-368 Supplier StatLab Supplier No. SSSLDLBLV
Add to Cart
Add to Cart
Zebra compatible xylene resistant slide labels.
  • 0.96875 x 875 in., 5.2 mil thick
  • 1-across, 1 in. core
  • 1,500 labels/roll
  • Perforated, gloss finish, opaque liner
Specifications
Product Type Slide Labels