Learn More
Sigma Aldrich Fine Chemicals Biosciences StainTray slide staining s1EA
Catalog No. 502906165
Encompass
These robust hunidity chambers offer a more user friendly approach to immunohistochemistry staining. This tray is suitable for routine staining requiring a humidity chamber. Wells between rails holds water securely without splashing. The plastic rails are covered with a polymer strip for perfectly holding slides even if tray is held at an angle. Rails are raised not only to avoid water touching the slides but to make them more easily retrievable. The base will also excess stain SOLUTIONon dripping from the slides.
Catalog No. 50-290-6165 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6701461EA
These robust hunidity chambers offer a more user friendly approach to immunohistochemistry staining. This tray is suitable for routine staining requiring a humidity chamber. Wells between rails holds water securely without splashing. The plastic rails are covered with a polymer strip for perfectly holding slides even if tray is held at an angle. Rails are raised not only to avoid water touching the slides but to make them more easily retrievable. The base will also excess stain SOLUTIONon dripping from the slides.