Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stak-a-Cab™ Stackable Cabinets
SCI_ED

Catalog No. S51012YSMF Shop All SciMatCo Products
Encompass_Preferred
Change view
Click to view available options
Color:
Gray
Yellow
Product Type:
Floor Stand
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
S51012YSMF Yellow Floor Stand
S51012GSMF Gray -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S51012YSMF Supplier SciMatCo Supplier No. SC1861/EMD

missing translation for 'validMainframeMsg'

Includes

Each part number represent one side-by-side cabinet; set of three cabinets shown represents three separate items stacked with base.

Use individually or stack up to three high with the optional floor stand.

No interior metal parts to corrode: wooden hinges and handles, polypropylene lining on cabinet floor, and polypropylene padlock assembly.

  • Lock with two keys and piano-style door hinges
  • All hardware is corrosion resistant
  • Liquid-tight trough floors
  • 10 x 10 in. (25 x 25cm) Hazard labels on both doors
  • Epoxy paint finish inside and out

Corrosives Cabinet

  • Capacity: 22 x 2.5L (approx. 14.5 gal./55L)
  • Blue finish
  • Accepts nitric acid compartment

Flammables Cabinet

  • Capacity: 16 x 1 gal. (approx. 60.5L)
  • Yellow finish

Combination Cabinet

  • Combination corrosives/flammables cabinet
  • Corrosives compartment capacity: 10 x 2.5L (approx. 6.5 gal./25L)
  • Flammables compartment capacity: 8 x 1 gal. (approx. 30L)
  • Gray finish

Floor Stands

  • Height: 4.5 in. (11cm)
  • Epoxy paint finish

Order Info

Interior: 15.125D x 45W x 16 in.H (40 x 114 x 41cm); Exterior: 18L x 47W x 18 in.H (46 x 119 x 46cm)

Compliance

Meet all applicable OSHA and NFPA standards.

Specifications

Surface Coating Epoxy
For Use With (Equipment) Flammables Stak-a-Cab cabinet
Width (English) Exterior 46.5 in.
Height (Metric) Exterior 11.43 cm
Includes Four built in adjustable leveling feet
Color Yellow
Height (English) Exterior 4.5 in.
Product Type Floor Stand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.