Learn More
MSC Tingley Magnaprene Jacket, Neoprene On Nylon, Waterproof, Large, 1 Ea.
Catalog No. 5000687122
Encompass
The Magnaprene jacket features a durable, single-coated neoprene on nylon construction designed for high performance and toughness in demanding environments. This jacket is 100% waterproof and provides excellent resistance to oils, acids, chemicals, and solvents.
  • Features 100% waterproof protection with stitched and taped seams
  • Flame resistant material tested per ASTM D6413; self-extinguishing with flameout and afterglow of ≤ 2 seconds
  • Generously cut with raglan sleeves for enhanced freedom of movement
  • Equipped with a storm fly front to seal out wind, rain, and contaminants
  • Includes elastic inner cuffs and adjustable take-up snaps on sleeves
  • Designed with a corduroy tip collar for neck comfort
  • Mildew resistant for long-lasting durability
Catalog No. 50-006-87122 Supplier MSC Supplier No. 35116532
The Magnaprene jacket features a durable, single-coated neoprene on nylon construction designed for high performance and toughness in demanding environments. This jacket is 100% waterproof and provides excellent resistance to oils, acids, chemicals, and solvents.
  • Features 100% waterproof protection with stitched and taped seams
  • Flame resistant material tested per ASTM D6413; self-extinguishing with flameout and afterglow of ≤ 2 seconds
  • Generously cut with raglan sleeves for enhanced freedom of movement
  • Equipped with a storm fly front to seal out wind, rain, and contaminants
  • Includes elastic inner cuffs and adjustable take-up snaps on sleeves
  • Designed with a corduroy tip collar for neck comfort
  • Mildew resistant for long-lasting durability