Learn More
Aviva Systems Biology Corp STAMBP Antibody - C-terminal region (ARP53527_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53527_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAMBP. It was validated on Western Blot. . Alternate names: AMSH, MGC126516, MGC126518ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-845