Learn More
Aviva Systems Biology Corp STAMBPL1 Antibody - N-terminal region (ARP57453_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57453_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAMBPL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STAMBPL1. Alternate names: ALMalpha, AMSH-FP, AMSH-LP, FLJ31524, KIAA1373, bA399O19.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62135