Learn More
MSC Minuteman Floor Stripper, 55 gal Drum, Multi-Surface
Catalog No. 502839109
Encompass
This floor stripper is designed for industrial use, offering effective removal on various surfaces. It comes in a large 55-gallon drum, making it suitable for extensive cleaning operations across different material types.
  • Stripper product type
  • Drum container type
  • 55 gallon container size
  • Applicable on concrete, terrazzo, vinyl, and vinyl composite tile (VCT)
  • Item is hazardous and ships via truck
Catalog No. 50-283-9109 Supplier MSC Supplier No. 73061749
This floor stripper is designed for industrial use, offering effective removal on various surfaces. It comes in a large 55-gallon drum, making it suitable for extensive cleaning operations across different material types.
  • Stripper product type
  • Drum container type
  • 55 gallon container size
  • Applicable on concrete, terrazzo, vinyl, and vinyl composite tile (VCT)
  • Item is hazardous and ships via truck