Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stampidine | 217178-62-6 | 100.0% | 25 MG
SDP

Catalog No. 5000231257
Encompass_Preferred

Stampidine is a nucleoside reverse transcriptase inhibitor (NRTI) used in research for its potent, broad-spectrum anti-HIV activity; it inhibits laboratory and clinical HIV-1 isolates at low nanomolar IC50s.

  • Potent anti-HIV activity with IC50s in the low nanomolar range.
  • Molecular formula: C20H23BrN3O8P.
  • Molecular weight: 544.29 g/mol.
  • Purity: 99.96% (as supplied).
  • Form: white to off-white solid powder.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Pack size: 25 mg.

Catalog No. 50-002-31257 Supplier Medchemexpress LLC Supplier No. HY12247025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stampidine is a nucleoside reverse transcriptase inhibitor (NRTI) used in research for its potent, broad-spectrum anti-HIV activity; it inhibits laboratory and clinical HIV-1 isolates at low nanomolar IC50s.

  • Potent anti-HIV activity with IC50s in the low nanomolar range.
  • Molecular formula: C20H23BrN3O8P.
  • Molecular weight: 544.29 g/mol.
  • Purity: 99.96% (as supplied).
  • Form: white to off-white solid powder.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Pack size: 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.