Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Cholesterol Standard
SDP

Catalog No. SB1012030 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB-1012-030 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SB-1012-030 Supplier Stanbio™ Supplier No. 1012030

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in chemical/clinical testing.

  • Concentration: 200mg/dL
  • Volume: 30mL

Specifications

Concentration 0.2 g/dL
Type Cholesterol Standard
Intended Use Chemical/clincal testing
Quantity 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.