Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Hema-Screen™ Lab Pack
SDP

Catalog No. 22022521 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-022-521 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-022-521 Supplier Stanbio™ Supplier No. 1290100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to collect specimens and perform assay in laboratory

  • Provides results in 2 minutes

For in-vitro diagnostic use only

Specifications

Content And Storage Room Temperature
Type Hema Screen™ Lab Pack
For Use With (Application) For in-vitro diagnostic use only
Includes Hema Screen™ developing solution, patient instructions, Wooden applicator and technical insert
Quantity 100 Tests
Test Time 2 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.