Learn More
Stanbio™ Quicktest RPR
SDP
Catalog No. SB1170100 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
100 Tests
3 x 5 mL
500 Tests
Type:
QuickTest RPR Test
QuickTest RPR Test Control
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
SB-1170-100 100 Tests QuickTest RPR Test
SB-1175-305 3 x 5 mL QuickTest RPR Test Control
SB-1170-500 500 Tests QuickTest RPR Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. SB-1170-100 Supplier Stanbio™ Supplier No. 1170100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
RPR antigen suspension, cardiolipin suspension, RPR reactive control, RPR weakly-reactive control, RPR non-reactive control
Carbon flocculation slide test
  • For qualitative and quantitative determination of reagin antibodies in serum or plasma
  • Macroscopic non-treponemal flocculation test that is used to detect and quantify reagin, an anti-lipoidal antibody found in serum or plasma from persons with syphilis and occasionally in persons with acute or chronic infections or conditions other than syphilis
  • Antigen is modification of VDRL antigen which contains microparticulate charcoal to enhance the visual difference between a reactive and non-reactive result
Specifications
Detectable Analytes Reagin (Syphilis)
Type QuickTest RPR Test
Includes RPR Antigen suspension, cardiolipin suspension, RPR reactive control, RPR weakly-reactive control, RPR non-reactive control
Quantity 100 Tests