Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ QuStick™ Strep A Kit
SDP

Catalog No. 22022516 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
25 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-022-516 50 Tests
22-022-515 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-022-516 Supplier Stanbio™ Supplier No. 6000050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Test strips, reagents, controls, throat swabs, extraction tubes, instructions

For diagnostic use

  • Sensitivity: 97% compared to culture
  • Results in 5 minutes

Certifications

CLIA waived

Specifications

Content And Storage Room Temperature
Clia Complexity Waived
Detectable Analytes Streptococci (Group A) Antigen
Sample Type Throat Swab
Sensitivity 97%
Type Streptococcus A Test Kit
Includes Test strip, reagents, control, throat swabs, extraction tubes and instructions
Quantity 50 Tests
Test Time 0.5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.